Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAD4 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303224100UL
Description
MAD4 Polyclonal specifically detects MAD4 in Mouse samples. It is validated for Western Blot.Specifications
MAD4 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
BHLHC12, bHLHc12Max-interacting transcriptional repressor MAD4, Class C basic helix-loop-helix protein 12, Mad4 homolog, MAD4Max-associated protein 4, MAX dimerization protein 4, Max dimerizer 4, MST149, MSTP149, MXD4 MAX dimerization protein 4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 110-209 of human MAD4 (NP_006445.1). | |
100 μL | |
Cell Cycle and Replication | |
10608 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction