Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAdCAM-1 Antibody (RM0284-11A55), Alexa Fluor™ 405, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211785AF405
Description
MAdCAM-1 Monoclonal specifically detects MAdCAM-1 in Mouse samples. It is validated for Western Blot.Specifications
MAdCAM-1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MADCAM1 | |
Mouse recombinant MAdCAM-1 (extracellular domain) | |
0.1 mL | |
8174 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0284-11A55 | |
Western Blot | |
hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1 | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction