Learn More
Madgetech™ RTD Probes Available on GSA/VA Contract for Federal Government customers only.
Precision 100 ohm, 4-wire RTD probes
Manufacturer:  MadgeTech IncSupplier Diversity Partner RTDSMS43/16636
 View more versions of this product
Catalog No. 15-060-056
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
6 in. Stainless Steel sheath, Metal strain relief wrapped
RTDTemp 101, RTDTemp 110, QuadRTD, OctRTD, RFRTDTemp101A Dataloggers
-328° to 1112°F
RTD Probe
Stainless Steel
-200° to 600°C