Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAF Mouse anti-Human, Clone: OTI2B12, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA812376

Description
MAF acts as a transcriptional activator or repressor. The protein is involved in embryonic lens fiber cell development.Specifications
MAF | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O75444 | |
MAF | |
Full length human recombit protein of human MAF produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI2B12 | |
Unconjugated | |
MAF | |
AYGRP, CCA4, CTRCT21, c-MAF | |
Mouse | |
Affinity Chromatography | |
RUO | |
4094 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title