Learn More
Aviva Systems Biology Corp MAFA Antibody - N-terminal region (ARP47760_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47760_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAFA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAFA. Alternate names: RIPE3b1, hMafAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-59660