Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova Human MAFB Partial ORF (NP_005452.2, 1 a.a. - 53 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00009935Q01S
Description
The protein encoded by this gene is a basic leucine zipper (bZIP) transcription factor that plays an important role in the regulation of lineage-specific hematopoiesis. The encoded nuclear protein represses ETS1-mediated transcription of erythroid-specific genes in myeloid cells. This gene contains no introns. [provided by RefSeq]
Sequence: MAAELSMGPELPTSPLAMEYVNDFDLLKFDVKKEPLGRAERPGRPCTRLQPAGSpecifications
NP_005452.2 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.57kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAELSMGPELPTSPLAMEYVNDFDLLKFDVKKEPLGRAERPGRPCTRLQPAG | |
RUO | |
MAFB | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
9935 | |
MAFB (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KRML|MGC43127 | |
MAFB | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title