Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MafK Antibody (2F7), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222570AF350
Description
MafK Monoclonal specifically detects MafK in Human samples. It is validated for Western Blot, ELISA.Specifications
MafK | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
MAFK | |
Recombinant human MAFK (1-156aa) purified from E. coli | |
0.1 mL | |
Primary | |
Human | |
IgG2a κ |
Western Blot, ELISA | |
2F7 | |
Western Blot, ELISA | |
FLJ32205, MGC71717, transcription factor MafK, ubiquitous (p18), v-maf avian musculoaponeurotic fibrosarcoma oncogene family, protein K, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene family, protein K, v-maf musculoaponeurotic fibrosarcoma oncogene homolog K (avian) | |
Mouse | |
Protein A purified | |
DNA replication Transcription Translation and Splicing, Epigenetics | |
7975.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction