Learn More
CPC Scientific H-Met-Glu-Val-Asp-Pro-Ile-Gly-His-Leu-Tyr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CANS006A
Encompass
Antigenic peptide encoded by the MAGE-3 gene that exhibits the strongest binding to HLA B44 and stimulates blood T lymphocytes that recognize tumor cells expressing MAGE-3, thereby a promising specific immunization of HLA-B44.ONE-LETTER SEQUENCE: MEVDPIGHLYMOLECULAR FORMULA: C53H80N12O16S1MOLECULAR WEIGHT:1173.36STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [178243-46-4], SYNONYMS: Melanoma-Associated Antigen 3 (167-176) (human)RESEARCH AREA: CancerREFERENCES: J. Herman et al., Immunogenetics, 43, 377 (1996); M.P. Velders et al., Crit. Rev. Immunol., 18, 7 (1998)
Catalog No. 50-210-8908
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.