Learn More
Abnova™ MAGEA11 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00004110T02
Catalog No. 89-951-427
Add to cart
Description
Description
Specifications
Specifications
293T
ENST00000370417
pCMV-MAGEA11 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
MAGEA11 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 35.5kDa
Transient overexpression cell lysate was tested with anti-MAGEA11 antibody (Mfr. No. H00004110-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human