Learn More
MAGEA11 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 154741AP
Catalog No. 50-173-0257
Add to cart
Description
Description
MAGEA11 is a member of the MAGEA gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 80% sequence identity between each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are expressed at a high level in a number of tumors of various histologic types, and are silent in normal tissues with the exception of testis and placenta. The MAGEA genes are clustered on chromosome Xq28. They may be implicated in some hereditary disorders, such as dyskeratosis congenita.
Specifications
Specifications
MAGEA11
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P43364
Cancer/testis antigen 1.11, CT1.11, MAGE 11, MAGE 11 antigen, MAGE11, MAGEA 11, MAGEA11, melanoma antigen family A, 11, Melanoma associated antigen 11
Rabbit
IgG
150 μL
-20°C
Primary
4110
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
MAGEA11
Liquid
MAGEA11
MAGEA11 Fusion Protein Ag7778
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human