Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAGEA12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567682
Description
MAGEA12 Polyclonal Antibody for Western Blot
Not known, though may play a role tumor transformation or progression. In vitro promotes cell viability in melanoma cell lines.
Specifications
MAGEA12 | |
Polyclonal | |
Unconjugated | |
MAGEA12 | |
Cancer/testis antigen 1.12; cancer/testis antigen family 1, member 12; CT1.12; MAGE family member A12; MAGE12; MAGE-12 antigen; MAGE12F antigen; MAGEA12; melanoma antigen family A, 12; melanoma antigen family A12; melanoma-associated antigen 12 | |
Rabbit | |
Affinity chromatography | |
RUO | |
4111 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P43365 | |
MAGEA12 | |
Synthesized peptide derived from human MAGEA12. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction