Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEA3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
$152.22 - $493.00
Specifications
Antigen | MAGEA3 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15640420
|
Novus Biologicals
NBP15640420UL |
20 μL |
Each for $152.22
|
|
NBP156404
|
Novus Biologicals
NBP156404 |
100 μL |
Each for $493.00
|
|
Description
MAGEA3 Polyclonal specifically detects MAGEA3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MAGEA3 | |
Polyclonal | |
Rabbit | |
Cancer | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Antigen MZ2-D, Cancer/testis antigen 1.3, CT1.3 MAGE-3 antigen, HIP8, MAGE3 MAGEA6, melanoma antigen family A, 3, melanoma-associated antigen 3, member 3, MGC14613 | |
MAGEA3 | |
IgG | |
Affinity Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
P43357 | |
4102 | |
Synthetic peptides corresponding to MAGEA3(melanoma antigen family A, 3) The peptide sequence was selected from the middle region of MAGEA3. Peptide sequence APEEKIWEELSVLEVFEGREDSILGDPKKLLTQHFVQENYLEYRQVPGSD. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title