Learn More
MAGEA4 Mouse anti-Human, Clone: OTI1F9, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA505362
Catalog No. 50-167-5850
Add to cart
Description
Description
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. At least four variants encoding the same protein have been found for this gene.
Specifications
Specifications
MAGEA4
Monoclonal
Unconjugated
MAGEA4
Liquid
MAGEA4
Full length human recombit protein of human MAGEA4 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI1F9
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P43358
CT1.4, MAGE-41, MAGE-X2, MAGE4, MAGE4A, MAGE4B
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4103