Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEA4, Mouse anti-Human, DyLight 405, Clone: CPTC-MAGEA4-1, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279861V
Description
MAGEA4 Monoclonal antibody specifically detects MAGEA4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
MAGEA4 | |
CPTC-MAGEA4-1 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
MAGEA4 | |
Recombinant full-length human MAGEA4 protein | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAGE-X2 antigen, melanoma antigen family A, 4, melanoma-associated antigen 4, member 4 | |
Mouse | |
IgG2c Kappa | |
0.1 mL | |
Cancer | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title