Learn More
Abnova Human MAGEA5 Partial ORF (NP_066387.1, 60 a.a. - 124 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00004104Q01S
Description
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. [provided by RefSeq]
Sequence: GPLKSPQGASAIPTAIDFTLWRQSIKGSSNQEEEGPSTSPDPESVFRAALSKKVADLIHFLLLKYSpecifications
NP_066387.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.89kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
GPLKSPQGASAIPTAIDFTLWRQSIKGSSNQEEEGPSTSPDPESVFRAALSKKVADLIHFLLLKY | |
RUO | |
MAGEA5 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
4104 | |
MAGEA5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAGE5|MAGEA4|MGC129526 | |
MAGEA5 | |
Yes |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.