Learn More
MAGEB4 Mouse anti-Human, Clone: OTI3H2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809147
Catalog No. 50-166-9304
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEB genes are clustered on chromosome Xp22-p21. This gene sequence ends in the first intron of MAGEB1, another family member. This gene is expressed in testis.
Specifications
Specifications
MAGEB4
Monoclonal
Unconjugated
MAGEB4
Lyophilized
MAGEB4
Human recombit protein fragment corresponding to amino acids 1-193 of human MAGEB4 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI3H2
PBS with 8% trehalose and no preservative; pH 7.3
O15481
CT3.6
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4115