Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGED2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Specifications

Antigen MAGED2
Immunogen Synthetic peptides corresponding to MAGED2 (melanoma antigen family D, 2) The peptide sequence was selected from the N terminal of MAGED2. Peptide sequence TEVSKTPEAREAPATQASSTTQLTDTQVLAAENKSLAADTKKQNADPQAV.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP168962 View Documents Novus BiologicalsSupplier Diversity Partner
NBP168962
100 ul This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

MAGED2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

MAGED2
Rabbit
Polyclonal
Unconjugated
10916
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to MAGED2 (melanoma antigen family D, 2) The peptide sequence was selected from the N terminal of MAGED2. Peptide sequence TEVSKTPEAREAPATQASSTTQLTDTQVLAAENKSLAADTKKQNADPQAV.
Primary
IgG
11B6MAGE-D2 antigen, BCG-1, BCG1MAGEDHCA10, breast cancer associated gene 1, Breast cancer-associated gene 1 protein, hepatocellular carcinoma associated protein, hepatocellular carcinoma-associated protein HCA10, Hepatocellular carcinoma-associated protein JCL-1, JCL-1, MAGE-D2, melanoma antigen family D, 2, melanoma-associated antigen D2, MGC8386
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit