Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAGEE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551808
Description
Immunogen sequence: STLFSSSASV DRNPSKCSLV LPSPRVTKAS VDSDSEGPKG AEGPIEFEVL RDCESPNSIS IMGLNTSRVA ITLKPQDPME QNVAELLQFL LVKDQSKYPI RE Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 60%.
This gene encodes an alpha-dystrobrevin-associated MAGE (melanoma-associated antigen) protein, which is a member of the MAGE family. The protein contains a nuclear localization signal in the N-terminus, 30 12-amino acid repeats beginning at nt 60 with the consensus sequence ASEGPSTSVLPT, and two MAGE domains in the C-terminus. It may play a signaling role in brain, muscle, and peripheral nerve. This gene is located on X chromosome in a region containing loci linked to cognitive disability.
Specifications
MAGEE1 | |
Polyclonal | |
Unconjugated | |
MAGEE1 | |
AI847422; Alpha-dystrobrevin-associated MAGE Protein; DAMAGE; dystrobrevin-associated MAGE protein; HCA1; hepatocellular carcinoma-associated HCA1; hepatocellular carcinoma-associated protein 1; hepatocellular carcinoma-associated protein HCA1; KIAA1587; MAGE family member E1; Magee1; MAGE-E1 antigen; melanoma antigen family E, 1; melanoma antigen family E1; melanoma antigen, family E, 1; melanoma-associated antigen E1; mMage-e1; RGD1560259 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57692 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCI5 | |
MAGEE1 | |
Recombinant protein corresponding to Human MAGEE1. Recombinant protein control fragment (Product #RP-92258). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction