Learn More
MAGEF1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 241051AP
Catalog No. 50-173-4238
Add to cart
Description
Description
Melanoma-associated antigen (MAGE) are completely silent in normal tissues, with the exception of male germ cells, and, for some of them, placenta. These antigens ought to be strictly tumor specific, expressed in tumor cells of various histological types. Because of their specific expression on tumor cells, these antigens are of particular interest for antitumor immunotherapy. Genes of the MAGE family direct the expression of tumor antigens that are recognized on a human melanoma by autologous cytolytic T lymphocytes. Though the function of MAGE is unknown, may play a role in embryonal development and tumor transformation or aspects of tumor progression.
Specifications
Specifications
MAGEF1
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HAY2
MAGE F1 antigen, MAGEF1, melanoma antigen family F, 1, Melanoma associated antigen F1
Rabbit
IgG
150 μL
-20°C
Primary
64110
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MAGEF1
Liquid
MAGEF1
MAGEF1 Fusion Protein Ag21147
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human