Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549843

Catalog No. PA549843


Add to cart

Description

Description

The antibody detects endogenous levels of total MAGI2 protein.

Seems to act as scaffold molecule at synaptic junctions by assembling neurotransmitter receptors and cell adhesion proteins. May play a role in regulating activin-mediated signaling in neuronal cells. Enhances the ability of PTEN to suppress AKT1 activation. Plays a role in nerve growth factor (NGF)-induced recruitment of RAPGEF2 to late endosomes and neurite outgrowth. Florindo C.S., Submitted (JUL-2003) to the EMBL/GenBank/DDBJ databases. The MGC Project Team, Genome Res. 14:2121-2127(2004). Devroe E., J. Biol. Chem. 279:24444-24451(2004).
Specifications

Specifications

MAGI2
Polyclonal
Unconjugated
MAGI2
Liquid
MAGI2
Synthesized peptide derived from internal of human MAGI2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q86UL8, Q9WVQ1, O88382
ACVRINP1, AIP1, KIAA0705
Rabbit
IgG
100 μL
-20°C
Primary
113970, 50791, 9863
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit