Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magid™ ROC™ PU50 Nylon Gloves

$71.00

Specifications

For Use With (Application) lint free applications
Glove Cuff Style Knit
Color White
Material Nylon
Sterility Non-sterile
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-047-297
View Documents
Magid™
PU508
8
Dozen of 12 for $71.00
Add to cart
 
19-047-298
View Documents
Magid™
PU509
9
Dozen of 12 for $71.00
Add to cart
 
19-047-299
View Documents
Magid™
PU5010
10
Dozen of 12 for $71.00
Add to cart
 
19-047-296
View Documents
Magid™
PU507
7
Dozen of 12 for $71.00
Add to cart
 
Description

Description

Specifications

Specifications

lint free applications
White
Non-sterile
Knit
Nylon
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit