Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Magill Catheter Forceps, Sklar, 9-3/4 in., Fenestrated, Straight, Serrated, Adult, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21289
Econo Magill Catheter Forceps, Sklar, 9-3/4 in., Fenestrated, Straight, Serrated, Adult, Stainless Steel. Econo Magill Catheter Forceps are used for grasping endotracheal tube during intubation and removing foreign bodies from the airway. They can also be used to grasp other types of catheters during insertion. Econo instruments are disposable, floor-grade instruments sold in bulk quantities.
Catalog No. 50-143-7586