Learn More
Roche Diagnostics MAGNA PURE LC PROCES CARTRIDGE  DFS Item
Manufacturer:  Roche Diagnostics 03004147001
Cartridge, Processing; Roche; Used as reaction vials for nucleic acid isolation using the MagNA Pure LC Instrument; Each cartridge contains 64 (8 x 8) separate vials; 160 cartridges/pack
Catalog No. 50-720-3010