Learn More
Roche Diagnostics MAGNA PURE LC RXN TIPS (LARGE)  DFS Item
Manufacturer:  Roche Diagnostics 03004171001
Tips, Reaction; Roche; Used in the MagNA Pure LC Instrument to dispense reagents and for magnetic-bead separation; Has integrated filters; Pipetting vol. range: 50-1000uL; Large; 32 tips per tray; 30 trays/pk.; 960 tips/pack
Catalog No. 50-720-3012