Learn More
Roche Diagnostics MAGNA PURE LC SAMPLE CARTRIDGE  DFS Item
Manufacturer:  Roche Diagnostics 03004112001
Cartridge, Sample; Roche; For MagNA Pure LC; For aliquoting primary sample, eluting nucleic acids from magnetic beads; Contains 32 (4 x 8) separate vials, vol. 1.5mL; Compatible w/std. 8-well multichannel pipettes; 120 cartridges/pk.
Catalog No. 50-720-3009