Learn More
Roche Diagnostics MAGNA PURE LC TIP STANDS  DFS Item
Manufacturer:  Roche Diagnostics 03004155001
Stand, Tip; Roche; Used for the temporary storage of tips for multiple use during the isolation process of the MagNA Pure LC Instrument; Decreases the number of required tips; 200 tip stands/pack
Catalog No. 50-720-3011