Learn More
Roche Diagnostics MAGNA PURE LC TUBS (LARGE)  DFS Item
Manufacturer:  Roche Diagnostics 03004040001
Tub, Reagent; Roche; For storing up to 100mL of MagNA Pure LC reagents on the stage of the MagNA Pure LC instrument; Can be sealed with MagNA Pure LC tub lids; Large; 120 tubs/pack
Catalog No. 50-720-3003