Learn More
Promega Plate Clamp 96
Recommended for automated protocols. Designed to ensure that nonskirted PCR plates are uniformly flat for liquid transfer on a robotic platform.
$297.00 - $518.00
Specifications
Content And Storage 15°C to 30°C
Product Type Accessory, Plate
Quantity 1/Ea.
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
PRV8251
View Documents
Promega
V8251
Plate Clamp 96
Each for $518.00
Add to cart
PRV8261
View Documents
Promega
V8261
Plate Stand
Each for $297.00
Add to cart
Description
Description
  • Plate clamp is designed to ensure that nonskirted PCR plates are uniformly flat for liquid transfer on a robotic platform
  • Plate Stand positions the Plate Clamp 96 assembly on the deck of a robotic workstation
Specifications
Specifications
15°C to 30°C
Accessory, Plate
1/Ea.