Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Magnesium 1,1,1-trifluoro-2,4-pentanedionate dihydrate, 98%

Click to view available options
Quantity:
2g
10g
240131-46-8
C10H12F6MgO6
366.495
MFCD00077561
OWJDZQKGGDMWCY-UDVCPWNYSA-L
magnesium trifluoroacetylacetonate dihydrate,magnesium1,1,1-trifluoro-2,4-pentanedionate,magnesium 1,1,1-trifluoro-2,4-pentanedionate 2-hydrate,3z-1,1,1-trifluoro-4-2z-5,5,5-trifluoro-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one dihydrate
91872989
magnesium;(Z)-5,5,5-trifluoro-4-oxopent-2-en-2-olate;dihydrate
CC(=CC(=O)C(F)(F)F)[O-].CC(=CC(=O)C(F)(F)F)[O-].O.O.[Mg+2]
240131-46-8
C10H12F6MgO6
366.495
MFCD00077561
OWJDZQKGGDMWCY-UDVCPWNYSA-L
magnesium trifluoroacetylacetonate dihydrate,magnesium1,1,1-trifluoro-2,4-pentanedionate,magnesium 1,1,1-trifluoro-2,4-pentanedionate 2-hydrate,3z-1,1,1-trifluoro-4-2z-5,5,5-trifluoro-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one dihydrate
91872989
magnesium;(Z)-5,5,5-trifluoro-4-oxopent-2-en-2-olate;dihydrate
CC(=CC(=O)C(F)(F)F)[O-].CC(=CC(=O)C(F)(F)F)[O-].O.O.[Mg+2]
Quantity 2g
Formula Weight 366.50 (330.47 Anhydrous)
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Magnesium 1,1,1-trifluoro-2,4-pentanedionate dihydrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning