Alfa Aesar™ Magnesium 2,4-pentanedionate hydrate, typically 98+% (metals basis)

Manufacturer: Alfa Aesar™ 1253218

Quantity 50g

 View more versions of this product

Catalog No. AA1253218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14024-56-7
    Formula Weight 222.52 (Anhydrous)
    MDL Number MFCD00150168
    Synonym Bis(acetylacetonato)magnesium; Bis(2,4-pentanedionato)magnesium
    Chemical Name or Material Magnesium 2,4-pentanedionate hydrate

    Specifications

    Formula Weight 222.52 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    MDL Number MFCD00150168
    Synonym Bis(acetylacetonato)magnesium; Bis(2,4-pentanedionato)magnesium
    Color White
    CAS 14024-56-7
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 260°C (dec.)
    Quantity 50g
    Chemical Name or Material Magnesium 2,4-pentanedionate hydrate
    Assay ≥98% (typically), (metals basis)

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit