Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium, 99.9+%, (trace metal basis), turnings, Thermo Scientific™

Click to view available options
Quantity:
500g
2.5kg
10kg
7439-95-4
Mg
24.305
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
5462224
CHEBI:25107
magnesium
[Mg]
7439-95-4
Mg
24.305
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
5462224
CHEBI:25107
magnesium
[Mg]
For ultra trace analysis
Flash Point 500°C
Quantity 500g
Density 1.74g/mL
Assay Percent Range 99.9+%
Linear Formula Mg
Fieser 01,627; 02,254; 03,189; 04,315; 05,419; 06,351; 07,218; 10,251; 12,290; 13,170; 15,120; 16,198
Packaging Plastic bottle
Formula Weight 24.3
Physical Form Turnings
Percent Purity ≥99.9%
Grade Trace Metal Basis
Total Trace Metal Impurities 0.10% max. (metals sought for: Si, Mn, Cu, Al, Fe, Pb, Ni, Sn)
Color Undesignated
Melting Point 651.0°C
Boiling Point 1107.0°C
Chemical Name or Material Magnesium, 99.9+%
Show More Show Less

GHS H Statement:
In contact with water releases flammable gases.
Self-heating in large quantities; may catch fire.
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 231-104-6

TSCA : TSCA

RUO – Research Use Only