Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium, 99+%, Acros Organics™

Click to view available options
Quantity:
25g
100g
500g
7439-95-4
Mg
24.305
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesio,sheet,powdered,magnesio italian,turnings,rieke's active,ribbon,unii-i38zp9992a,hsdb 654,nitrate, acs
5462224
CHEBI:25107
magnesium
[Mg]
7439-95-4
Mg
24.305
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesio,sheet,powdered,magnesio italian,turnings,rieke's active,ribbon,unii-i38zp9992a,hsdb 654,nitrate, acs
5462224
CHEBI:25107
magnesium
[Mg]
Quantity 25g
Linear Formula Mg
Solubility Information Solubility in water: insoluble.
Formula Weight 24.3
Percent Purity ≥99%
Grade Pure
Melting Point 651.0°C
Boiling Point 1107.0°C
Chemical Name or Material Magnesium

GHS H Statement In contact with water releases flammable gases which may ignite spontaneously. Catches fire spontaneously if exposed to air.

GHS P Statement Keep away from any possible contact with water, because of violent reaction and possible flash fire. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection.

Danger