Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Acetate, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

16674-78-5
C4H14MgO8
214.453
XKPKPGCRSHFTKM-UHFFFAOYSA-L
134717

High quality chemicals for laboratory and specialized industrial use

CAS 16674-78-5
Molecular Formula C4H14MgO8
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Mg+2]
IUPAC Name magnesium;diacetate;tetrahydrate
Molecular Weight (g/mol) 214.453
PubChem CID 134717
Formula Weight 214.45
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.45kg
Quantity 500g
Chemical Name or Material Magnesium Acetate, 4-Hydrate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less