Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium Acetate Tetrahydrate, ACS, 98-102%, Alfa Aesar™
Click to view available options
Quantity:
100g
500g
2.5kg
Specifications
Assay Percent Range | 98.0-102.0% |
Beilstein | 3730605 |
Appearance | White crystals |
Solubility Information | Soluble in water. |
Formula Weight | 214.45 (142.39anhydrous) |
Physical Form | Crystals |
Grade | ACS |
Density | 1.454 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
H500
EINECSNumber : 205-554-9
TSCA : Yes
Recommended Storage : Ambient temperatures