Alfa Aesar™ Magnesium aluminum oxide, 99.9% (metals basis excluding Ca), Ca <25ppm

Manufacturer: Alfa Aesar™ 3638822

Quantity 100g

 View more versions of this product

Catalog No. AA3638822

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 12068-51-8
    Formula Weight 142.26
    MDL Number MFCD00049475
    Chemical Name or Material Magnesium aluminum oxide

    Specifications

    Formula Weight 142.26
    Odor Odorless
    Particle Size 3 to 12mm
    Physical Form Fused Pieces
    MDL Number MFCD00049475
    CAS 12068-51-8
    TSCA TSCA
    Quantity 100g
    Chemical Name or Material Magnesium aluminum oxide
    Assay 99.9%, (metals basis excluding Ca), Ca <25ppm

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit