Alfa Aesar™ Magnesium aluminum oxide, 99% (metals basis)

Manufacturer: Alfa Aesar™ 2295036

Quantity 500g

 View more versions of this product

Catalog No. AA2295036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12068-51-8
    Formula Weight 142.26
    MDL Number MFCD00049475
    Synonym Magnesium aluminate
    Chemical Name or Material Magnesium aluminum oxide

    Specifications

    Formula Weight 142.26
    Mesh Size −325 Mesh
    MDL Number MFCD00049475
    Odor Odorless
    Physical Form Powder
    Synonym Magnesium aluminate
    TSCA TSCA
    CAS 12068-51-8
    Quantity 500g
    Chemical Name or Material Magnesium aluminum oxide
    Assay 99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit