Alfa Aesar™ Magnesium Aluminum Zinc wire, 3.18mm (0.125 in.) dia., 90cm (35 in.) long

Manufacturer: Alfa Aesar™ 14092RN

Quantity 5 x 90cm

 View more versions of this product

Catalog No. AA14092RN

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    MDL Number MFCD01074901
    Chemical Name or Material Magnesium Aluminum Zinc wire, 3.18mm (0.125 in.) dia., 90cm (35 in.) long

    Specifications

    MDL Number MFCD01074901
    Weight ≈12.44g/90cm
    Quantity 5 x 90cm
    TSCA TSCA
    Physical Form Wire
    Odor Odorless
    Avg. Mol. Wt. or Mol. Wt. Range Mg:Al:Zn; 93:6:1 wt%
    Chemical Name or Material Magnesium Aluminum Zinc wire, 3.18mm (0.125 in.) dia., 90cm (35 in.) long

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit