Alfa Aesar™ Magnesium aluminum zirconium oxide, 99% (metals basis excluding Ca) Ca 0.1-1%

Manufacturer: Alfa Aesar™ 36368-22

Quantity 100g

 View more versions of this product

Catalog No. AA3636822

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 53169-11-2
    Formula Weight 265.46
    MDL Number MFCD00171346
    Chemical Name or Material Magnesium aluminum zirconium oxide

    Specifications

    Formula Weight 265.46
    Particle Size 3 to 12mm
    MDL Number MFCD00171346
    Physical Form Sintered Lump
    CAS 53169-11-2
    Quantity 100g
    Chemical Name or Material Magnesium aluminum zirconium oxide
    Assay 99%, (metals basis excluding Ca) Ca 0.1 to 1%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit