Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium bis(diisopropylamide), 0.7M solution in THF, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
AcroSeal™ Glass bottle
23293-23-4,109-99-9,513-35-9
C12H30MgN2
226.691
MFCD03093019
GOVRLZMVMIIFQC-UHFFFAOYSA-N
magnesium-bis diisopropyl amide,
87510612
magnesium;N-propan-2-ylpropan-2-amine
CC(C)NC(C)C.CC(C)NC(C)C.[Mg]
23293-23-4,109-99-9,513-35-9
C12H30MgN2
226.691
MFCD03093019
GOVRLZMVMIIFQC-UHFFFAOYSA-N
magnesium-bis diisopropyl amide,
87510612
magnesium;N-propan-2-ylpropan-2-amine
CC(C)NC(C)C.CC(C)NC(C)C.[Mg]
Appearance of Solution Yellowish-brown solution
Concentration 0.6 to 0.8M
Density 0.8800g/mL
Formula Weight 224.68
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: vigorous reaction to mg(oh)2, diisopropylamine.
Specific Gravity 0.88
Boiling Point 66.0°C
Quantity 100mL
Flash Point −45°C
Chemical Name or Material Magnesium bis(diisopropylamide), 0.7M solution in THF
Show More Show Less

GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Harmful if swallowed. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Suspected of causing cancer. Suspected of causing genetic defects. Harmful to aquatic life with long lasting effects. Reacts violently with water. May form explosive peroxides.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger