Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium bromide ethyl etherate, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
29858-07-9
C4H10Br2MgO
258.24
MFCD00064500
JGZKUKYUQJUUNE-UHFFFAOYSA-L
magnesium bromide diethyl etherate, magnesium bromide etherate, magnesium bromide ethyl etherate, grignard reagent
11010650
magnesium;ethoxyethane;dibromide
CCOCC.[Mg+2].[Br-].[Br-]
29858-07-9
C4H10Br2MgO
258.24
MFCD00064500
JGZKUKYUQJUUNE-UHFFFAOYSA-L
magnesium bromide diethyl etherate, magnesium bromide etherate, magnesium bromide ethyl etherate, grignard reagent
11010650
magnesium;ethoxyethane;dibromide
CCOCC.[Mg+2].[Br-].[Br-]
Flash Point 35°C
Color Beige to White
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula (C2H5)2O·MgBr2
Fieser 01,629; 07,218; 13,259; 17,174
Solubility Information Solubility in water: soluble
Formula Weight 258.24
Physical Form Crystalline Powder/Chunks
Percent Purity 99%
Chemical Name or Material Magnesium bromide ethyl etherate
Show More Show Less

GHS H Statement
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning