Alfa Aesar™ Magnesium bromide hexahydrate, 98+%

Manufacturer: Alfa Aesar™ 46120A3

Quantity 2kg

 View more versions of this product

Catalog No. AA46120A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13446-53-2
    Molecular Formula Br2Mg·6H2O
    Linear Formula MgBr2·6H2O
    Formula Weight 292.22 (184.13 Anhydrous)
    MDL Number MFCD00149780
    Chemical Name or Material Magnesium bromide hexahydrate

    Specifications

    Formula Weight 292.22 (184.13 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula Br2Mg·6H2O
    Solubility Information Soluble in water, somewhat soluble in alcohol
    Health Hazard 1 Warning
    TSCA TSCA
    Odor Odorless
    Linear Formula MgBr2·6H2O
    MDL Number MFCD00149780
    Color Colorless
    Melting Point 172°C
    Physical Form Crystalline
    Sensitivity Hygroscopic
    CAS 13446-53-2
    Quantity 2kg
    Density 2.00g/mL
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Magnesium bromide hexahydrate
    Assay ≥98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit