Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium bromide hexahydrate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Glass bottle
13446-53-2
Br2Mg·6H2O
292.21
MFCD00149780
LGLXXNHIGIJYQQ-UHFFFAOYSA-L
acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent
17749097
magnesium;dibromide;hexahydrate
O.O.O.O.O.O.[Mg+2].[Br-].[Br-]
13446-53-2
Br2Mg·6H2O
292.21
MFCD00149780
LGLXXNHIGIJYQQ-UHFFFAOYSA-L
acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent
17749097
magnesium;dibromide;hexahydrate
O.O.O.O.O.O.[Mg+2].[Br-].[Br-]
Assay Percent Range 99% min. (Complexometry)
Linear Formula MgBr2·6H2O
Fieser 13,259; 15,194; 16,199
Merck Index 14, 5659
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 292.21
Physical Form Crystalline Powder
Percent Purity 99+%
Grade Analysis
Cadmium (Cd) 50ppm max.
Chloride 500ppm max
Chlorine (Cl) 500ppm max.
Copper (Cu) 50ppm max.
Iron (Fe) 50ppm max.
Lead (Pb) 50ppm max.
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in alcohol
Sulfate 200ppm max.
Trace Analysis 50ppm max.
Color White
Melting Point 165.0°C
pH 6.5 to 8 (5% soln. at 20°C)
Quantity 1kg
Chemical Name or Material Magnesium bromide hexahydrate, p.a.
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning