Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium bromide hexahydrate, extra pure, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
13446-53-2
Br2Mg·6H2O
292.21
MFCD00149780
LGLXXNHIGIJYQQ-UHFFFAOYSA-L
acmc-1bp1b,magnesium bromide-water 1/2/6,magnesium 2+ ion hexahydrate dibromide,magnesium 2+ hexahydrate dibromide
17749097
magnesium;dibromide;hexahydrate
O.O.O.O.O.O.[Mg+2].[Br-].[Br-]
13446-53-2
Br2Mg·6H2O
292.21
MFCD00149780
LGLXXNHIGIJYQQ-UHFFFAOYSA-L
acmc-1bp1b,magnesium bromide-water 1/2/6,magnesium 2+ ion hexahydrate dibromide,magnesium 2+ hexahydrate dibromide
17749097
magnesium;dibromide;hexahydrate
O.O.O.O.O.O.[Mg+2].[Br-].[Br-]
Linear Formula MgBr2·6H2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 292.21
Grade Extra Pure
Packaging Plastic bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in alcohol
Melting Point 165.0°C
Quantity 1kg
Chemical Name or Material Magnesium bromide hexahydrate

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning