Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium carbonate basic, light, for analysis, 40.0-45.0% Magnesium oxide, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
Packaging:
Plastic drum
39409-82-0
1/4H2O2Mg·5/4H2O
121.41
MFCD07371358
39409-82-0
1/4H2O2Mg·5/4H2O
121.41
MFCD07371358
Linear Formula Mg(OH)2·5H2O
Merck Index 15, 5724
Arsenic (As) 2ppm max.
Solubility Information Solubility in water: practically insoluble
Formula Weight 121.41
Physical Form Powder
Percent Purity 40.0 to 45.0% (MgO)
Grade Analysis
Calcium (Ca) 7500ppm max.
Chloride 700ppm max
Chlorine (Cl) 700ppm max.
Density 2.1600g/mL
Heavy Metals (as Pb) 20ppm max.
Insoluble Matter 0.05% max. (in acetic acid)
Iron (Fe) 400ppm max.
Packaging Plastic drum
Specific Gravity 2.16
Sulfate 3000ppm max.
Water Soluble Substances 1% max.
Color White
Quantity 1kg
Chemical Name or Material Magnesium carbonate basic, light
Show More Show Less