Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium carbonate hydroxide tetrahydrate, White powder, MgO approx.40-43.5%, Alfa Aesar™
Click to view available options
Quantity:
100g
500g
Specifications
Assay Percent Range | MgO approx.40-43.5% |
Solubility Information | Slightly soluble in water. |
Formula Weight | 467.67 (395.58 Anhydrous) |
Physical Form | -325 Mesh Powder |
Grade | Reagent |
Odor | Odorless |
Anhydrous or Hydrate | Tetrahydrate |
Color | White |
Quantity | 500g |
Chemical Name or Material | Magnesium carbonate hydroxide tetrahydrate |
Safety and Handling
EINECSNumber : 235-192-7
RTECSNumber : FG1797000
TSCA : Yes
Recommended Storage : Ambient temperatures