Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium Chloride, 51% (w/v), Ricca Chemical
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
Specifications
Color | Colorless |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Aqueous Solution |
Quantity | 500 mL |
Physical Form | Liquid |
Concentration or Composition Notes | 51% (w/v) |
Chemical Name or Material | Magnesium Chloride Solution |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction