Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride, 6-Hydrate, Crystal, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7791-18-6
MgCl2·6H2O
CAS 7791-18-6
Molecular Formula MgCl2·6H2O
Formula Weight 203.3
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.57kg
Quantity 2.5kg
Chemical Name or Material Magnesium Chloride, 6-Hydrate
Grade FCC/USP