Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride, 6-Hydrate, Crystal, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

7791-18-6
MgCl2·6H2O

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Molecular Biology Applications
Formula Weight 203.3
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.57kg
Quantity 500g
Chemical Name or Material Magnesium Chloride, 6-Hydrate
Grade Ultrapure Bioreagent