Magnesium chloride, anhydrous, 99%, Alfa Aesar™

For Use As a Catalyst, Flocculating Agent, Various Magnesium Cements

Manufacturer: Alfa Aesar 12315C4

Quantity 25kg

 View more versions of this product

Catalog No. AA12315C4

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7786-30-3
    Physical Form Crystals
    Assay Percent Range 99%
    Molecular Formula MgCl2
    Formula Weight 95.22
    MDL Number MFCD00011106
    Chemical Name or Material Magnesium Chloride, Anhydrous

    Specifications

    Physical Form Crystals
    Formula Weight 95.22
    Quantity 25kg
    Appearance White to Yellow Crystals
    Color White to yellow
    EINECS Number 232-094-6
    Molecular Formula MgCl2
    MDL Number MFCD00011106
    Odor Odorless
    Assay Percent Range 99%
    CAS 7786-30-3
    Chemical Name or Material Magnesium Chloride, Anhydrous

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit