Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium chloride hexahydrate, 98+%, extra pure, Acros Organics

Click to view available options
Quantity:
1kg
5kg
7791-18-6
Cl2Mg·6H2O
203.31
7791-18-6
Cl2Mg·6H2O
203.31
Assay Percent Range 98.0 to 101% (Complexometry)
Linear Formula MgCl2·6H2O
Merck Index 15,5726
Solubility Information Solubility in water: 2350g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 203.31
Percent Purity 98+%
Grade Extra Pure
Packaging Plastic bottle
Solubility Solubility in water: 2350g/L (20°C). Other solubilities: soluble in alcohol
Melting Point 117.0°C
Quantity 1kg
Chemical Name or Material Magnesium chloride hexahydrate
Show More Show Less